FisherBiotech™ Spacers for FB-GC10-1 Vertical Gel Caster
Spacers for use with FB-GC10-1 Vertical Gel Caster.
Manufacturer:  FisherBiotech™ P7SC
 View more versions of this product
Catalog No. FB-VE10-34
Specifications Description & Specifications
Specifications
5.9 in.
Two side spacers and one bottom spacer
15cm
3.9 in.
0.05cm
10cm
0.8cm
0.003 in.
10cm
3.9 in.
Owl™ P7-CST multiple gradient gel caster
0.008cm
Spacer Set
10 x 10cm
0.03 in.