Please login to your online account to display your discounted pricing
FisherBiotech™ Horizontal Minigel Systems: Combs
For use with Horizontal Minigel systems
Manufacturer: fisherbiotech™  B1A5C
 View more versions of this product
Catalog No. FB-SB-733
  • / Each
Description & Specifications
Specifications
Teeth Count 5
For Use With (Equipment) Owl™ EasyCast™ B1A mini gel and A2-OK multi gel electrophoresis system
Volume (Fill) Metric 55μL
Width (Metric) Teeth 11.3mm
Thickness (Metric) Teeth 1mm
Product Type Standard Comb