Please login to your online account to display your discounted pricing

FisherBiotech™ Horizontal Electrophoresis Systems


Choose from four systems

Manufacturer: fisherbiotech™  FIS2318

 View more versions of this product

Catalog No. FBSB2318

  • / Each
 

Description & Specifications

Specifications

Dimensions (L x W x H) Exterior 13 x 11 x 5 in. (33 x 27 x 13cm)
Gel Dimensions 18 x 23cm
Includes Buffer chamber, safety lid with attached power supply Leads, UV transparent gel tray, nonskid Rubber feet, two 1.5mm thick combs (12 and 20-well)
Model Wide format system
Sample Size 25 to 250
Type Standard
Volume (Metric) Buffer 1.2L