missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FisherBiotech™ Gel Casting Pouches for FB-GC16-1 Vertical Gel Casters

$35.68 - $85.56

Specifications

For Use With (Equipment) FB-GC16-1 Vertical Gel Casters
Length (Metric) 16cm
Product Type Gel Casting Pouch
Width (Metric) 14cm
Gel Dimensions 16 x 14cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FB-GC16-11 FisherBiotech™
GP225
25/Pk. Pack of 25 Each for $35.68
FB-GC16-12 FisherBiotech™
GP2100
100/Pk. Pack of 100 Each for $85.56
Specifications Description & Specifications

Specifications

FB-GC16-1 Vertical Gel Casters
Gel Casting Pouch
16 x 14cm
16cm
14cm
5.5 in.