FisherBiotech™ FB-VE10-1 Vertical Electrophoresis System: Combs
For use with FB-VE10-1 vertical electrophoresis systems
Manufacturer:  FisherBiotech™ MP12A
 View more versions of this product
Catalog No. FB-VE10-25
Specifications Description & Specifications
Specifications
12
34μL
Comb
0.43cm
0.05cm
Owl™ P8 series electrophoresis system
0.16 in.
10 x 10cm