Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
Supplier: Fisherbrand™ MP20A
Specifications
Owl P8 Series Electrophoresis System | |
10 x 10 cm | |
0.05 cm | |
5.9 in. | |
15 cm | |
3.9 in. | |
13 cm | |
0.16 cm | |
13 cm |
Comb | |
20 | |
13 μL | |
3.9 in. | |
10 cm | |
0.06 in. | |
10 cm | |
5.11 in. |