FisherBiotech™ DNA Sequencing Systems
Run samples in all gel lanes without loss of data or questionable interpretation of the banding pattern
$1,371.18 - $2,428.79
Includes: Main chamber with aluminum backplate, clamps, removable lower buffer chamber, two notched glass plates, two blank glass plates, two sharktooth combs, one well comb, two spacer sets
Requires: High-voltage power supply
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
FB-SEQ-2045 FisherBiotech™
S4SXMFIS
Each for $1,371.18
FB-SEQ-3545 FisherBiotech™
S3SXMFIS
Each for $2,428.79
Specifications
Description
  • Integral anodized aluminum backplate distributes heat evenly across the entire gel surface to minimize band “smiling”
  • Screw clamps are mounted directly on the unit to ensure leakproof seal between gel sandwich and upper buffer chamber
  • Removable waste bottle allows for easy buffer disposal
  • Interlocking lids on upper and lower chambers provide safety