Please login to your online account to display your discounted pricing
FisherBiotech™ 8-Tooth Combs for Horizontal Midigel Systems
8-Tooth Combs for Horizontal Midigel Systems
$121.04 - $121.04
Specifications
For Use With (Equipment) Owl™ A1 large gel electrophoresis system
Product Type Standard Comb
Teeth Count 8
Width (English) Teeth 0.54 in.
Width (Metric) Teeth 1.39cm
View More Specs
Products
Catalog Number Mfr. No. Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity    
FB-SB-1325
fisherbiotech™
A18D
0.15cm 55μL Each for $121.04
FB-SB-1324
fisherbiotech™
A1-8C
0.1cm 65μL Each for $121.04
Description & Specifications
Specifications
For Use With (Equipment) Owl™ A1 large gel electrophoresis system
Product Type Standard Comb
Teeth Count 8
Width (English) Teeth 0.54 in.
Width (Metric) Teeth 1.39cm
  • Accessory for use with Horizontal Midigel Systems