Please login to your online account to display your discounted pricing
FisherBiotech™ 24-Tooth Combs for Horizontal Midigel Systems
24-Tooth Combs for Horizontal Midigel Systems
Manufacturer: fisherbiotech™  A124D
 View more versions of this product
Catalog No. FB-SB-1333
  • / Each
Description & Specifications
Specifications
Teeth Count 24
Product Type Standard Comb
For Use With (Equipment) Owl™ A1 large gel electrophoresis system
Width (English) Teeth 0.13 in.
Volume (Fill) Metric 25μL
Width (Metric) Teeth 0.35cm
Thickness (Metric) Teeth 0.15cm