Please login to your online account to display your discounted pricing
FisherBiotech™ 16-Tooth Combs for Horizontal Midigel Systems
16-Tooth Combs for Horizontal Midigel Systems
Manufacturer: fisherbiotech™  A116C
 View more versions of this product
Catalog No. FB-SB-1328
  • / Each
Description & Specifications
Specifications
Teeth Count 16
Product Type Standard Comb
For Use With (Equipment) Owl™ A1 large gel electrophoresis system
Width (English) Teeth 0.24 in.
Volume (Fill) Metric 30μL
Width (Metric) Teeth 0.61cm
Thickness (Metric) Teeth 0.1cm