FisherBiotech™ 12-Tooth Combs for Horizontal Midigel Systems
12-Tooth Combs for Horizontal Midigel Systems
$111.25 - $121.04
Specifications
For Use With (Equipment) Owl™ A1 large gel electrophoresis system
Product Type Standard Comb
Teeth Count 12
Width (English) Teeth 0.34 in.
Width (Metric) Teeth 0.87cm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity  
Catalog Number Mfr. No. Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity  
FB-SB-1327 FisherBiotech™
A112D
0.15cm 60μL Each for $111.25
FB-SB-1326 FisherBiotech™
A112C
0.1cm 40μL Each for $121.04
Specifications
Description
  • Accessory for use with Horizontal Midigel Systems
Description & Specifications
Specifications
Owl™ A1 large gel electrophoresis system
12
0.87cm
Standard Comb
0.34 in.