Please login to your online account to display your discounted pricing
FisherBiotech™ 12-Tooth Combs for Horizontal Midigel Systems
12-Tooth Combs for Horizontal Midigel Systems
Manufacturer: fisherbiotech™  A112C
 View more versions of this product
Catalog No. FB-SB-1326
  • / Each
Description & Specifications
Specifications
Teeth Count 12
Product Type Standard Comb
Volume (Fill) Metric 40μL
For Use With (Equipment) Owl™ A1 large gel electrophoresis system
Width (English) Teeth 0.34 in.
Width (Metric) Teeth 0.87cm
Thickness (Metric) Teeth 0.1cm