Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
$884.31 - $954.40
Specifications
Includes Cap with tubing and liquid level sensors
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Product Type Price Qty  
Catalog Number Mfr. No. Capacity (Metric) Product Type Price Qty  
14430212 missing translation for 'viewDocuments' Fisherbrand™
MXN10810
4L Waste Bottle Each for $884.31
14430211 missing translation for 'viewDocuments' Fisherbrand™
MXN10809
2L Rinse Bottle Each for $954.40
14430210 missing translation for 'viewDocuments' Fisherbrand™
MXN10808
2L Wash Bottle B Each for $947.17
14430209 missing translation for 'viewDocuments' Fisherbrand™
MXN10807
2L Wash Bottle A Each for $947.17
Description
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Specifications
Specifications
Cap with tubing and liquid level sensors