Fisher Scientific™ Wave Motion Shaker
Heavy-duty rotator moves in a unique three-dimensional orbital wave motion.
Manufacturer:  Fisher Scientific™ BKL281111FEMD
 View more versions of this product
Catalog No. S06620
Specifications Description & Specifications
Specifications
Wave action shaker
20 lb.
33.02cm
14 in.
27.94cm
4° to 40°C
13 in.
14 x 14 x 11 in. (35.6 x 35.56 x 27.94cm)
6w
0.6A
10.8kg
11 in.
115V 50/60Hz
UL/CSA
35.56cm
50/60Hz
5 to 120rpm
0 to 12
14 in.
Up to 12 Hours or set for continuous
115V
24 lb.
35.6cm