missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Vortex Mixer Accessories: Foam Insert Set

For use with Fisherbrand vortex mixers.

Supplier:  Fisherbrand™ 80862994

 View more versions of this product

Catalog No. 02-215-394


Description

Description

For use with Fisherbrand vortex mixers.

Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.

Foam Insert Variety Pack

  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm

Specifications

Specifications

9 to 13 mm, 14 to 19 mm, 20 to 29 mm
Foam
9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Foam Insert Set
14 to 19 mm
SDS
Documents

Documents

Accessories
Promotions

Promotions