missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Visual Alarm Timers

Easy-to-use timers. Fisherbrand Visual Alarm Timers come with either an audible alarm or bright flashing LED visual alarm.

Manufacturer:  Fisherbrand™ 8196

 View more versions of this product

Catalog No. S90205B



Includes: Stand, clip, integrated magnet, two AAA batteries

Description

Description

  • Controlled with only three buttons
  • Automatic memory resets the display to the last programmed time
  • “Thru-zero” feature allows counting down, alarming and then counting up
  • 0.38 in. (9.6mm) digits are readable from 10 ft. (3m)
  • With stand, clip and integrated magnet
  • Dimensions: 3 × 2.25 × 0.75 in. (7.6 × 5.7 × 1.9cm)

99-Minute Model

  • Counts down from 99 minutes, 59 seconds in second increments
9999-Minute Model
  • Counts down from 9999 minutes in minute increments and has a countdown resolution of one second for times under 99 minutes
Specifications

Specifications

Flashing yellow LED visual and audible
9999 min.
LCD
Education
Two AAA batteries, integrated magnet, stand, clip
AAA
Visual Alarm Timer
Documents