Fisherbrand™ Visual Alarm Timers
Easy-to-use timers. Fisherbrand Visual Alarm Timers come with either an audible alarm or bright flashing LED visual alarm.
Manufacturer:  Fisherbrand™ 8195
 View more versions of this product
Catalog No. S90205A
Includes: Stand, clip, integrated magnet, two AAA batteries
Description
Description
  • Controlled with only three buttons
  • Automatic memory resets the display to the last programmed time
  • “Thru-zero” feature allows counting down, alarming and then counting up
  • 0.38 in. (9.6mm) digits are readable from 10 ft. (3m)
  • With stand, clip and integrated magnet
  • Dimensions: 3 × 2.25 × 0.75 in. (7.6 × 5.7 × 1.9cm)
99-Minute Model
  • Counts down from 99 minutes, 59 seconds in second increments
9999-Minute Model
  • Counts down from 9999 minutes in minute increments and has a countdown resolution of one second for times under 99 minutes
Specifications
Specifications
Flashing yellow LED visual and audible
99 min. 59 sec.
LCD
Education
Two AAA batteries, integrated magnet, stand, clip
AAA
Visual Alarm Timer
Documents