Please login to your online account to display your discounted pricing
Fisher Scientific™ Vial Blocks for Incubating/Cooling Mini Shakers
For holding vials of different sizes
Manufacturer: troemner llc  980FIVLB28
 View more versions of this product
Catalog No. 02-217-732
  • / Each
Description & Specifications
Specifications
Size 6 x 28mm
Type Vial Block
For Use With (Equipment) Incubating/cooling mini shaker
Material Solid anodized Aluminum
Wash/Waste Bottle Capacity 28mm