Please login to your online account to display your discounted pricing
Fisher Scientific™ Universal Spill Kit
Small enough to fit on a lab bench.
Manufacturer: fisher science education  630EMD*
 View more versions of this product
 Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Sorbent and pillows not suitable for use with hydrofluoric acid.
Catalog No. S47440
  • / Pack of 18 Each
Description & Specifications
Specifications
Capacity 0.22 gal., 0.83L
For Use With (Application) Safe for use on all chemicals except hydrofluoric acid
Capacity (English) 0.22 gal.
Capacity (Metric) 0.83L
Product Type Lab Pillow