Fisherbrand™ Ultrasonic Cleaner Perforated Pans
Supports easy removal of items from ultrasonic cleaners.
Manufacturer:  Fisherbrand™ CPN169061
 View more versions of this product
Catalog No. 15-335-110A
Description
Description
Perforated Pans
  • Support items in the tank for direct cleaning
  • For easy removal of items from the tank
Beaker Covers
  • Reduce evaporation and eliminate splashing problems
  • Beaker positioning covers required to suspend standard 600mL beakers in the tank
Beaker Rings
  • Allows placement of 250 or 400mL beakers within the standard beaker cover
† Discontinued model.
Specifications
Specifications
9.5L
FS110 ultrasonic cleaner
1216 oz., 9.5L
1216 oz.
Perforated Pan
Documents