missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Ultra Low Temperature Recorder

Programmable high and low alarms Features three LEDs (green, yellow and red) to alert user of log-time, temperature warning limit and temperature breach

Manufacturer:  Fisherbrand™ CRYO-TEMP-CERTX

 View more versions of this product

Catalog No. 15060002CER


Alert:

Fire, explosion, and severe burn hazard. Do not recharge,dissassembled, heat above 212°F, incinerate or expose contents to water.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Requires : Interface Package: 15-060-038. Utilizes a USB docking station (sold separately) to communicate with a personal computer.

Description

Description

  • Records temperatures as low as -123°F (-86°C)
  • Magnetic start
  • Thermistor Sensor
  • Provides date and time-stamped temperature readings
  • Data can be viewed in graphical or tabular form
  • Programmable high and low alarms Features three LEDs (green, yellow and red) to alert user of log-time, temperature warning limit and temperature breach
  • Material: ABS Plastic, rated IP65
  • Battery Life: Two years typical at 25°C, one minute reading intervals

Cryopreservations; Blood Plasma freezers; Vaccines; Cold chain; Low-temperature monitoring; Dry-ice shipment/storage; Shipping and storage applications

Certifications

NIST Traceable

Compliance

CE marked

Specifications

Specifications

CRYO-TEMPX
Digital calibration
LED Indicator
2 Years
0 to 100%
For cryopreservations, Blood plasma freezers, vaccines, cold chain, low temperature monitoring, dry ice shipment/storage, shipping and storage applications
0.01°C
-122.8° to 95°F
1.4 oz.
NIST calibration certificate
CE, NIST
3.6V Lithium
32767 Readings
Cryo-Temp Datalogger
ABS Plastic
-86° to 35°C
Thermistor
40g
Documents