missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Tubing Y Polypropylene Connectors

Work in any tubing or vacuum system.

Manufacturer:  Fisherbrand™ 222555EMD

 View more versions of this product

Catalog No. S50701B



Description

Description

  • Excellent chemical resistance
  • Polypropylene
  • Autoclavable
  •  Serrated connecting surfaces to prevent tubing slipping
  • Withstands flows to 20 p.s.i.
Specifications

Specifications

Transparent
0.18 to 0.32 in.
1.5 in.
Tubing Connector
Used for any tubing or vacuum system
5.33 mm
Y
0.8 in.
Autoclavable
Polypropylene
38.1 mm
4.5 to 8.1mm
20 psi
0.21 in.
20.3 mm
Documents