Please login to your online account to display your discounted pricing

Fisherbrand™ Tubing T Connectors

Work in any tubing or vacuum system.

$61.50 - $61.50

Specifications

Color Transparent
Autoclavable Yes
For Use With Used for any tubing or vacuum system
Material Polypropylene
Product Type Tubing Connector
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Inside Dimensions (L x W x H) Fits Tubing Price Quantity    

S50702A

 
fisherbrand™
227905EMD
0.12 to 0.15 in. 0.15 x 0.8 x 0.58 in. (3.8 x 20.32 x 14.7mm) 3 to 3.8mm Pack of 20 Each for $61.50
Description & Specifications

Specifications

Color Transparent
Autoclavable Yes
For Use With Used for any tubing or vacuum system
Material Polypropylene
Product Type Tubing Connector
Max. Pressure 20psi
Shape T

  • Excellent chemical and temperature resistance
  • Serrations hold tubing tightly and prevent slipping