missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Polypropylene Tubing Connectors and Adapters
Work in any tubing or vacuum system.
Manufacturer: Fisherbrand™ 227245EMD
Description
- Excellent chemical and temperature resistance
- Serrated connecting surfaces to prevent tubing slippage
- Autoclavable Polypropylene
- Adapters allow for connecting different diameter tubing
Specifications
Autoclavable | |
Polypropylene | |
36.8 mm | |
3 to 5.3mm | |
14.5 mm | |
Straight | |
20 psi |
0.125 to 0.03 in. | |
1.45 in. | |
Tubing Connector | |
Used for any tubing or vacuum system | |
0.57 in. | |
White |