missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Polypropylene Tubing Connectors and Adapters
Work in any tubing or vacuum system.
Manufacturer: Fisherbrand™ 227545
Description
- Excellent chemical and temperature resistance
- Serrated connecting surfaces to prevent tubing slippage
- Autoclavable Polypropylene
- Adapters allow for connecting different diameter tubing
Specifications
Autoclavable | |
Polypropylene | |
83.82 mm | |
3 to 7.8mm | |
20.5 mm | |
Straight-stepped | |
20 psi |
0.125 to 0.312 in. to 0.438 to 0.625 in. | |
3.3 in. | |
Tubing Connector | |
Used for any tubing or vacuum system | |
0.81 in. | |
White |