Please login to your online account to display your discounted pricing
Fisher Scientific™ Tube Blocks for Incubating/Cooling Mini Shakers
For microtubes, test tubes and centrifuge tubes
Manufacturer: troemner llc  980FIMTBL0
 View more versions of this product
Catalog No. 02-217-715
  • / Each
Description & Specifications
Specifications
Size 30 x 0.5mL
Type Microtube Block
For Use With (Equipment) Incubating/cooling mini shaker
Material Solid anodized Aluminum
Wash/Waste Bottle Capacity 0.5mL