Please login to your online account to display your discounted pricing
Fisher Scientific™ Tube Blocks for Incubating/Cooling Mini Shakers
For microtubes, test tubes and centrifuge tubes
Manufacturer: troemner llc  980FISTB12
 View more versions of this product
Catalog No. 02-217-722
  • / Each
Description & Specifications
Specifications
Size 20 x 12 to 13mm
Type Standard test tube block
For Use With (Equipment) Incubating/cooling mini shaker
Material Solid anodized Aluminum
Wash/Waste Bottle Capacity 12-13mm