Fisherbrand™ Tube Blocks for Incubating/Cooling Mini Shakers
For microtubes, test tubes and centrifuge tubes.
Manufacturer:  Fisherbrand™ 980FISTB06
 View more versions of this product
Catalog No. 02-217-720
Description
Description
  • Modular blocks constructed from solid anodized aluminum block
  • Tube close to block walls allow for maximum temperature transfer
Specifications
Specifications
30 x 6mm
Incubating/cooling mini shaker
6mm
Standard test tube blockþStandard test tube block
Solid anodized Aluminum
Documents