Please login to your online account to display your discounted pricing
Fisher Scientific™ Tube Blocks for Incubating/Cooling Mini Shakers
For microtubes, test tubes and centrifuge tubes
Manufacturer: troemner llc  980FICTBL1
 View more versions of this product
Catalog No. 02-217-718
  • / Each
Description & Specifications
Specifications
Size 12 x 15mL
Type Centrifuge tube block
For Use With (Equipment) Incubating/cooling mini shaker
Material Solid anodized Aluminum
Wash/Waste Bottle Capacity 15mL