Fisherbrand™ Traceable™ Noncontact Infrared Thermometers
Portable thermometers measure infrared surface temperatures with view fields of 6:1 to 8:1
Manufacturer:  Fisherbrand™ CON4317
 View more versions of this product
Catalog No. 15-077-57
Description
Description
Portable thermometers measure infrared surface temperatures with view fields of 6:1 to 8:1. Fisher Scientific™ Traceable™ Noncontact Infrared Thermometers are ideal for petroleum, electronics, plant and field use.
If a unit has a view field of 7:1 and the thermometer is held 14 in. (35.6cm) from object, it reads infrared radiation emitting from a 2 in. dia. (5.1cm) spot.
  • Switch to view either Celsius or Fahrenheit scales Laser sighting beam permits aiming precisely at the target surface
  • 15-077-57 and 06-664-39 assure accurate measurements using adjustable control to account for differences in surface infrared emissivity of objects
  • One-second sampling, backlighting button, data hold, low-battery indicator and dual ranges
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Specifications
−4° to 752°F
752°F
−4°F
ISO 17025, A2LA, NIST
Carrying case, 9-volt battery
197mm
9.62 oz.
±3°C or ±3%
−20° to 400°C
400°C
−20°C
Petroleum, electronics, plant and field use
7.75 in.
0.1°
0.23kg
Digital LCD
Documents
Accessories