Fisherbrand™ Traceable™ Logger-Trac™ Temperature Datalogger Accessories
Accessories for Traceable™ Logger-Trac™ Temperature Datalogger
Manufacturer:  Fisherbrand™ CON6545
 View more versions of this product
Catalog No. 15-081-145
Description
Description
Accessories for Traceable™ Logger-Trac™ Temperature Datalogger
15-081-145
  • Waterproof paper sleeves
  • For protecting temperature probe sensor
  • Transparent
15-081-148
  • USB standard A to Mini B, IC-9P or USB to serial converter
  • USB Version 2.0, ELV and RoHS compliant
  • Cable length: 1.5m
  • Multiple cradles allow for batch programming up to 8 units
Specifications
Specifications
Waterproof paper sleeves
Protecting probe sensor
Fisherbrand Traceable Logger-Trac Temperature Datalogger
Documents