missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Traceable™ Jumbo Refrigerator/Freezer Thermometers
Perfect for monitoring freezers, water baths, heating blocks, incubators and refrigerators
Supplier: Fisherbrand™ 9000896
Includes: Unit is supplied with a Traceable Certificate, flip-open stand for lab bench, slot for wall mounting, one AAA alkaline battery and Velcro™ and magnetic strips to attach to any surface
Description
Perfect for monitoring freezers, water baths, heating blocks, incubators and refrigerators. Fisher Scientific™ Traceable™ Jumbo Refrigerator/Freezer Thermometer accurately monitors temperature with an enclosed “temperature-buffered”sensor.
- Bottle insulates sealed sensor from rapid temperature changes when refrigerator door is opened
- Thermometer fulfills all CDC thermometer and vaccine thermometer requirements
- Easy-view, jumbo 1.5 in. (38.1 cm) high digits may be read from 30 ft. (9m)
- Sensor and 10 ft. cable perform accurately even when both are under water
- High-impact, chemical-resistant ABS plastic case
- Triple display simultaneously shows maximum, minimum and current probe temperatures plus room temperature
- Min./max. monitors high/low readings overnight, on weekends or for any time period — a significant advantage over “current reading-only” glass thermometers
- Range is -58° to +158°F and -50° to +70°C with a resolution of 0.01° and accuracy of ±1°C
- Alarm feature provides alert when temperature rises above or falls below a setpoint
- Alarm is programmable in 1° increments
- 10 ft. (3m) micro-cable permits refrigerator doors to close on it
- Solid-state probes eliminate mercury contamination in refrigerators
- Sealed probe is housed in a miniature bottle filled with nontoxic glycol; solution is GRAS (Generally Recognized as Safe)
- Eliminates concerns about incidental contact with food or drinking water
Order Info
- Case: 3.87 x 4.37 x 0.87 in. (9.83 x 11.1 x 2.2cm)
- Sealed bottle probe measures 1 x 2.5 in. (2.5 x 6.4cm)
- Naked sensor is 0.18 dia. x 0.75 in. long (4.6 x 19mm)
- Replacement battery (02-440-3A) available
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to +standards provided by NIST™. | |
Monitoring freezers, water baths, heating blocks, incubators and refrigerators | |
11.1 cm | |
120 in. | |
304.8 cm | |
0.1°C | |
110 g | |
2.5 in. | |
70°C | |
-50°C | |
-50°C to +70°C | |
9.82 cm |
2.5 x 3.87 x 4.37 in. (6.4 x 9.82 x 11.1 cm) | |
4.37 in. | |
Sealed bottle probe, traceable certificate, flip open stand, wall mount, Velcro™, magnetic strips and AAA Alkaline battery | |
64 mm | |
ABS Plastic | |
4 oz. | |
3.87 in. | |
158°F | |
-58°F | |
-58°F to +158°F | |
±1°C |