missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ International Standards Extreme-Accuracy Digital Thermometer

Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.

Supplier:  Fisherbrand™ 9367594

 View more versions of this product

Catalog No. 15-078-181



Description

Description

Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.

Units read 0.00, 25.00, 37.00, or a combination of all three in °F and °C.

  • Provides accurate readings with tip penetration of 0.33 in.
  • Records time/date for min/max readings and hi/lo alarm alerts
  • Displays minimum/maximum and current temperatures
  • Min/max monitors readings over any period; displays exact time/date of min/max temperature
  • Alarm provides two visual LEDs and two audio alerts when temperature exceeds set points; displays exact time/date of dual thermometer alarms
  • Programmable in 0.1° increments; alarms signal continuously even if temperature returns to non-alarm range
  • Resolution: 0.01°
  • Accuracy: ±0.05°C within ±2°C of standard
  • Probe Dia.: 0.12 in. (0.30 cm)
  • Stem Length: 6.25 in. (15.87 cm)
  • Overall Length: 9 in. (22.86 cm)
  • Cable: 5 ft. (1.5 m)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Compliance

Meets ITS-90, International Temperature Standard.

Specifications

Specifications

ISO 17025, A2LA, NIST
3.2 mm
Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
1.9 cm
6.25 in.
15.9 cm
0.01°C
140 g
4.25 in.
70°C
-50°C
-50°C to +70°C
AA
LED
Reads to 0.00, 25.00, and 37.00
0.125 in.
4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9 cm)
4.25 in.
Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)
107 mm
ABS Plastic
5 oz.
2.75 in.
158°F
-58.00°F
-58° to +158°F
Water Baths
±0.05°C within ±2°C of tested standard points, ±0.3°C otherwise
6.9 cm
SDS
Documents

Documents

Promotions

Promotions