missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ International Standards Extreme-Accuracy Digital Thermometers

Ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater

Manufacturer:  Fisherbrand™ CON4241

 View more versions of this product

Catalog No. 15-078-178



Includes: Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)

Description

Description

Ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater. Fisher Scientific™ Traceable™ International Standards Extreme-Accuracy Digital Thermometers have an impervious stainless-steel probe that shows slightest temperature change in liquid, air/gas, and semisolids.

Units read 0.00, 25.00, 37.00, or a combination of all three in °F and °C.

  • Provides accurate readings with tip penetration of 0.33 in.
  • Records time/date for min/max readings and hi/lo alarm alerts
  • Displays minimum/maximum and current temperatures
  • Min/max monitors readings over any period; displays exact time/date of min/max temperature
  • Alarm provides two visual LEDs and two audio alerts when temperature exceeds set points; displays exact time/date of dual thermometer alarms
  • Programmable in 0.1° increments; alarms signal continuously even if temperature returns to non-alarm range
  • Resolution: 0.01°
  • Accuracy: ±0.05°C within ±2°C of standard
  • Probe Dia.: 0.12 in. (0.30cm)
  • Stem Length: 6.25 in. (15.87cm)
  • Overall Length: 9 in. (22.86cm)
  • Cable: 5 ft. (1.5m)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Compliance

Meets ITS-90, International Temperature Standard.

Specifications

Specifications

28.4° to 35.6°F
35.6°F
28.4°F
ISO 17025, A2LA, NIST
4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9cm)
0.75 in.
Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring)
107mm
ABS Plastic
5 oz.
2.75 in.
±0.05°C
4.25 in.
−2.00° to 2.00°C
2°C
−2°C
0.12 in.
Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
1.9cm
6.25 in.
15.9cm
0.01°C
140g
6.9cm
LED
Documents