Please login to your online account to display your discounted pricing

Fisher Scientific™ Traceable™ Dual-Display Digital Stopwatch


Quartz-crystal design is accurate to 0.0005%—over 100 times more accurate than other stopwatches

Manufacturer: fisher scientific™  CON1034FN

 View more versions of this product

Stopwatch; lanyard

Catalog No. 02-261-953

  • / Each
 

Description & Specifications

Specifications

Accuracy 0.001%
For Use With (Application) Enzyme tests, viscosity measurements, animal studies and all timed lab tests
Height (English) Display 0.75 in.
Includes Stopwatch; lanyard
Length (English) 2.5 in.
Resolution 0.01 sec.
Weight (Metric) 0.07kg
Width (English) 3 in.
Certifications/Compliance ISO 17025, A2LA, NIST
Dimensions (L x W x H) 2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9cm)
Display Type LCD
Height (English) 0.75 in.
Height (Metric) Display 1.9cm
Height (Metric) 1.9cm
Length (Metric) 6.35cm
Material ABS Plastic
Weight (English) 2.5 oz.
Width (Metric) 7.62cm
Product Type Dual-Display Digital Stopwatch
Time Range 10 hrs.

Engineered for ease of operation, accuracy, and reliability. Fisher Scientific™ Traceable™ Dual-Display Digital Stopwatches provide precise, Traceable timing for enzyme tests, viscosity measurements, animal studies, and all timed lab tests. They are made for scientists in industrial, educational, hospital, and governmental labs. Quartz-crystal design is accurate to 0.0005%—over 100 times more accurate than other stopwatches.

Set countdown time from 10 hours to 1 second. When zero is reached, timer alarms and repeats countdown from the programmed time. Repeat cycle automatically continues until stopped. Display count box shows how many times the cycle has been repeated. LCD screen continually displays time remaining to zero. 

  • Quartz-crystal design is accurate to 0.0005%—over 100 times more accurate than other stopwatches
  • Functions include single-action, time-out, interval-split, cumulative split, continuous roll-over timing, and countdown alarm timing
  • Dual display shows cumulative and interval split
  • Features eight recallable memories, plus fastest and slowest lap times
  • Repeat timer provides repeat function of countdown alarm timing
  • Tough, water-resistant ABS plastic case seals out moisture, dust, and fumes encountered in harsh lab, plant, or field environments
  • Timing range is 9 hours, 59 minutes, 59 seconds, and 99 hundredths
  • Resolution is 0.01 second
  • Accuracy is 0.0005%
  • Size: 6.4 x 7.6 x 1.9cm (2.5 x 3 x 0.75 in.)
  • Weight: 2.5 oz.

Replacement battery: Cat. No. 02-440-1A

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.