Fisher Scientific™ Tissue Path™ Universal Modular Filing System
Stores 22% more slides or 32% more specimen blocks than standard units
$85.03 - $488.17
Specifications
For Use With (Equipment) Universal modular filing system
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
22-244-029 Fisher Scientific™
58193
Cabinet Each for $488.17
22-244-030 Fisher Scientific™
58199
Cabinet Each for $347.91
22-244-031 Fisher Scientific™
58194
MicroSlide™ Drawer Each for $85.03
22-244032 Fisher Scientific™
58195
CasSette/Block Drawer Pack of 2 Each for $115.00
22-244-035 Fisher Scientific™
58198
Drawer Each for $117.20
Specifications
Description
  • For microscope slides and histology cassettes
  • Made of extruded aluminum shells with a durable anodized finish
  • Slotted handle on each drawer for identification labeling
  • Interchangeable drawers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Universal modular filing system