Fisherbrand™ Tissue Path™ IV Biopsy Cassettes
Finely perforated to prevent loss of small biopsy specimens during processing
$366.97 - $373.16
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
22-313733
|
Fisherbrand™
58392 |
Yellow |
Case of 500 Each for $373.16
|
|
22-035411
|
Fisherbrand™
58454 |
White |
Case of 500 Each for $366.97
|
|
22-282255
|
Fisherbrand™
58452 |
Green |
Case of 500 Each for $366.97
|
|
22-282253
|
Fisherbrand™
58451 |
Gray |
Case of 500 Each for $366.97
|
|
22-313732
|
Fisherbrand™
58387 |
Powder Blue |
Case of 500 Each for $373.16
|
|
Description
- Same two-piece molded hinge design as Tissue Path IV Cassettes
- Three-sided writing surface assures positive identification
- Resistant to solvents, decalcifying solutions and compatible with microwave techniques
Order Info
- Packaged in two dispenser boxes, 250 per box
Specifications
Biopsy Cassette | |
Plastic |