Please login to your online account to display your discounted pricing
Fisher Scientific™ Tissue Path™ High-Profile Microtome Blades
Heat-treated, high carbon blades
$182.06 - $182.06
Products
Catalog Number Mfr. No. Price Quantity    
22030530
fisher scientific™
58054
Pack of 50 Each for $182.06
Description & Specifications
  • Precision-ground for optimal angle and facet configuration
  • Offers greater rigidity to prevent chattering and produces excellent ribboning characteristics
Packaged in easy-to-use dispenser packs.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.