Please login to your online account to display your discounted pricing

Fisher Scientific™ Technical Pens


Ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams.

Manufacturer: fisher scientific™  CON3047

 View more versions of this product

Catalog No. 13-383-1G

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Product Type Technical Pen
Width (Metric) Line 0.2mm
For Use With (Application) Ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams
Quantity 6 Pack
Color Green

Fisher Scientific™ Technical Pens are ideal for recording results in lab notebooks, fill in charts, draw graphs, mark recording paper and produce diagrams. 6 per pack.

Available in multiple colors and widths.