Fisherbrand™ Support Stands for Overhead Stirrerss
Provides stability for overhead stirrers.
Supplier:  Fisherbrand™ 370209049
 View more versions of this product
Catalog No. 14-512-909
Explore available promotions
Alert:
14-512-909 is NOT recommended for Models A30 or AP50R
Description
Description
Provides stability, reduces possible tipping and cuts vibration
  • Stainless steel base with two long front legs and one short back leg
  • Rubber pads on each foot
  • 29 in. overall height
Specifications
Specifications
0.62 in.
Overhead Stirrer
73.6 cm
1.58 cm
29 in.
Support Stand