Please login to your online account to display your discounted pricing
Fisher Scientific™ Standard Exhaust Oil Filters
As supplied with Fisher Scientific Maxima C Plus pumps
$321.44 - $804.04
Specifications
Type Standard Exhaust Oil Filter
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Height (English) Height (Metric) Price Quantity    
01-257-50
fisher scientific™
01-257-50
2.5 in. 6.35cm 4.62 in. 11.73cm Each for $507.23
01-257-5A
fisher scientific™
01-257-5A
2.5 in. 6.35cm 2.5 in. 6.35cm Each for $321.44
01-257-5C
fisher scientific™
01-257-5C
5 in. 12.7cm 6.25 in. 15.87cm Each for $804.04
Description & Specifications
Specifications
Type Standard Exhaust Oil Filter
These compact, round filters simply screw into threaded exhaust port to collect oil expelled during operation. A replaceable filter element within the oil filter separates the oil into particles no larger than 0.3μm, and drops collected oil back into the oil case.
  • Filter element is easily replaced
  • Reorder number located on filter canister
1For M4C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
2For M4C or M8C vacuum pumps designed after August 1999 (S/N FKK090000 and newer).