missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Stainless Steel Shafts for Mixing Impellers

Composed of 316L stainless steel and passivated for maximum corrosion protection. Available in multiple sizes.

Manufacturer:  Fisherbrand™ A753FSR

 View more versions of this product

Catalog No. 14-500-371



Description

Description

  • Composed of stainless steel 316L
  • .95 cm (0.375 in.) shafts have a turned down 0.8 cm (0.3125 in.) end
  • Passivated for corrosion resistance
  • Specifications

    Specifications

    0.37 in.
    Any propeller with a 0.95cm or 0.8cm dia. bore
    76.2 cm
    0.95 cm
    30 in.
    Shaft Accessory
    SDS
    Documents