Fisher Scientific™ Specific Gravity Hydrometers
Double scales can be read without turning hydrometer
Manufacturer:  Fisher Scientific™ 11510C
 View more versions of this product
Catalog No. 11-510C
Specifications Description & Specifications
Specifications
0.001
305mm
300mL Borosilicate Glass cylinder
250mL
ASTM E100, ASTM E126
Measure true specific gravity of liquids lighter than Water
0.895 to 1.01
Specific Gravity Hydrometer
Accessories