Fisher Scientific™ Specific Gravity Hydrometers
Double scales can be read without turning hydrometer
Manufacturer:  Fisher Scientific™ 11510BC
 View more versions of this product
Catalog No. 11-510BC
Specifications Description & Specifications
Specifications
0.001
310mm
300mL Borosilicate Glass cylinder
250mL
3-point Certification, ASTM E100, ASTM E126
Measure true specific gravity of liquids lighter than Water
0.795 to 0.91
Specific Gravity Hydrometer