Please login to your online account to display your discounted pricing

Fisher Scientific™ Specific Gravity Hydrometers


Double scales can be read without turning hydrometer

Manufacturer: fisher scientific™  11512CC

 View more versions of this product

Catalog No. 11-512CC

  • / Each
 

Description & Specifications

Specifications

Product Type Specific Gravity Hydrometer
Specific Gravity Range 0.89 to 1
Volume (Metric) 140mL
For Use With (Application) Measure true specific gravity of liquids lighter than Water
Increments 0.002
Length (Metric) 180mm
Certifications/Compliance 3-point Certification, ASTM E100, ASTM E126
For Use With (Equipment) 175mL Borosilicate Glass cylinder