Fisherbrand™ Specific Gravity Hydrometers
Measure true specific gravity of liquids.
Description
Measure true specific gravity of liquids.
Measure true specific gravity of liquids.
- Chemical-resistant bulbs and stems
- Streamlined to avoid trapping air bubbles
- With directional device that stops the instrument with the scale facing the user for more convenient, more accurate readings
Order Info
A certificate of calibration is available; simply add the letter “C” to the end of the catalog number when ordering.
Compliance
Meet or exceed ASTM™ E 100 for accuracy, ASTM E 126 for calibration
Specifications
Specific Gravity Hydrometer | |
ASTM E100, ASTM E126 | |
300mL Borosilicate Glass cylinder | |
Measure true specific gravity of liquids heavier than Water | |
140 mL |
0.005 | |
160 mm | |
Soda Lime Glass | |
1.200 to 1.425 |