Please login to your online account to display your discounted pricing
Fisher Scientific™ Slide Holders
Holders keep slides clean and untouched, and can be stored in sturdy steel cabinets with one or two drawers.
Manufacturer: fisher scientific™  CON1007EMD
 View more versions of this product
Catalog No. S14162
  • / Pack of 12 Each
Description & Specifications
Specifications
Device Holds 48 slides (3 x 5 in.)
Quantity 12/Pk. cards and 1 plastic box
Slide Holders
  • Add, remove or change cards for fast updating and filing; adapts to your slide ID system (numbers, letters, dates, names or color coding)
  • Visible, four-position tabs; ruled data lines keyed to numbered slots and allow complete slide identification
  • 3 × 5 in. (7.6 × 12.7cm) cardboard fitted with protective plastic frame accepts four slides of 75 × 25mm or 3 × 1 in. size of any thickness
One and two door cabinets sold separately (Cat. Nos. S94845 and S94846).