Fisherbrand™ Simultaneous Start Timer
Operates on all channels simultaneously. Fisherbrand Simultaneous Start Timer has four countdown channels, a stopwatch channel and a time-of-day channel.
Manufacturer:  Fisherbrand™ 8614
Catalog No. S90204
Description
Description
  • Countdown channels may be set independently and started simultaneously or individually
  • Counts both up and down to 99 hours, 59 minutes, 59 seconds
  • Unique “thru-zero” feature allows counting down, alarming and then counting up
  • Large, bright display
  • Extra-loud alarm
  • Contains an integrated flip-open stand, clip fastener and magnetic back
  • Dimensions: 2.75 × 2.75 × 0.75 in. (7.0 × 7.0 × 1.9cm)
Specifications
Specifications
Extra loud alarm
357 button cell batteries (2)
99 Hours 59 min. 59 sec.
Simultaneous Start Timer
Documents