Please login to your online account to display your discounted pricing

Fisher Scientific™ Replcement Tips for Model 505 and 705 Sonic Dismembrator


Replcement Tips for Model 505 and 705 Sonic Dismembrator

Manufacturer: fisher scientific™  4406

 View more versions of this product

Catalog No. FB4406

  • / Each
 

Description & Specifications

Specifications

Diameter (English) 0.5 in.
Type Replacement Tip
For Use With (Equipment) 0.5 in. probe of model 505 and 705 sonic dismembrator
Diameter (Metric) 1.27cm