Fisherbrand™ Replacement Tubing Set for Peristaltic Tubing Pumps
Provide fine resolution with a wide flow range used with food, pharmaceuticals and other critical solutions with this tubing. Fisherbrand™ Replacement Tubing for Peristaltic Tubing Pumps include silicone tubing and color-coded fittings.
Manufacturer:  Fisherbrand™ CON3371
 View more versions of this product
Catalog No. 15-079-607
Description
Description
Provide fine resolution with a wide flow range used with food, pharmaceuticals and other critical solutions with this tubing. Fisherbrand™ Replacement Tubing for Peristaltic Tubing Pumps include silicone tubing and color-coded fittings.
Specifications
Specifications
Yes
Ultra-Low flow, Low flow and Medium flow Variable-Speed Peristaltic Tubing Pumps
500°F
–62°C
Silicone
260°C
-80°F
1/50 in.
Documents